PYREX® Volumetric Flasks, Snap Cap, Class A, Corning
100 ml, 100, ±0.08 ml
Supplier: Corning
Número de proveedor #: 5580-100
PYREX® Volumetric Flasks, Snap Cap, Class A, Corning
Catalog # 89090-600
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
PYREX® glass, clear, with snap-caps.
- Heavy beads and tubing rings tooled for snap-cap fit
- Graduation line is sharp and permanent
- No snap caps for 1 and 2 ml sizes
Documents
Especificaciones
- Cap Size:100
- Capacity:100 ml
- Tolerance:±0.08 ml
- Cat. No.:89090-600
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact