KIMAX® Volumetric Flasks with Color-Coded [ST] PTFE Stopper, Class A, Kimble Chase
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificados
Acerca de este artículo
These flasks are calibrated to contain and feature a graduation ring blasted on the neck
Sizes 10mL and larger feature a marking area. Supplied with a [ST] PTFE stopper with color-coded handle. The 2mL size is shaped like a test tube; all other sizes are of a conventional flask shape. Sizes 5mL and larger are designed from ASTM Specification E288, Class A unserialized requirements.
Información relacionada
Frequently Bought Together