Chem Master Neoprene/Rubber Gloves, Showa
Catalog # 32892-059
Supplier: SHOWA-BEST GLOVE INC.
Product available on GSA Advantage®
Specifications
- Color del guante:Black
- Tamaño del guante:L
- Ambidiestro/Específico para cada mano:Específico para cada mano
- Longitud:33 cm (13")
- Grosor:26 mil.
- Tipo de puño:Puño recto
- Acabado:Tractor Tread
- Referencia:32892-059
- Nº de proveedor:CHML-09
Specifications
About this item
Hand and product protection for manufacturing, electronics, machine work, food processing, pharmaceutical, and laboratory applications.
- Resists a broad range of chemicals in many applications
- 100% cotton flock lining
- Provide comfort, flexibility, and tactility
- Self-flushing tractor tread grip for fluid runoff increases gripping effectiveness
Unsupported neoprene over natural rubber construction offers abrasion, tear, and puncture resistantance.
Color: black over blue.
Certifications: Sanitized gloves are made using only listed materials and levels outlined in 21 CFR 170-199.