AlphaTec® 09-928 Chemical resistant gloves, Ansell
Catalog # 32889-998
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Specifications
- Color del guante:Black
- Tamaño del guante:L
- Ambidiestro/Específico para cada mano:Específico para cada mano
- Desechable / Reutilizable:Reutilizable
- Descripción:Gauntlet
- Longitud:45.7 cm (17⅞")
- Tipo de puño:Puño tipo guante
- Acabado:Smooth
- Referencia:32889-998
- Modelo:9-928
- Nº de proveedor:213321
Specifications
About this item
Gloves are fully coated with reinforced neoprene coating to provide good all-around resistance to abrasions, cuts, and chemicals.
- Strong seams prevent hand irritation
- Good resistance to abrasions, cuts, and chemicals
- Natural fit
- Great comfort
Interlock knit cotton liner enhances puncture and snag resistance. Curved fingers and wing thumb provide natural fit and greater comfort. Two-piece lining eliminates seams from working areas to prevent hand irritation. Applications extend to refining and petrochemicals; handling oils, acids, caustics, and alcohols; degreasing; plating; and metal fabrication.