PYREX® Volumetric Flasks, Polyethylene Stopper, Class A, Corning
Supplier: Corning
PYREX® Volumetric Flasks, Polyethylene Stopper, Class A, Corning
Supplier: Corning
Certificates
Acerca de este artículo
PYREX® borosilicate glass, clear, with LDPE stoppers.
- Graduation line is sharp and permanent
- White marks and inscriptions are easy to read
Volumetric flasks is made with increased strength through machine blown bodies. Heavy tubing rings and heavy beaded seal is tooled for polyethylene tubing.
8 Options Available Below
Product Details & Documents
PYREX® borosilicate glass, clear, with LDPE stoppers.
- Graduation line is sharp and permanent
- White marks and inscriptions are easy to read
Volumetric flasks is made with increased strength through machine blown bodies. Heavy tubing rings and heavy beaded seal is tooled for polyethylene tubing.
Specifications for Product Family
Especificaciones
- Catalog No:
- 29619-198
- 89091-098
- 89090-602
- 89090-676
- 89090-710
- 89090-748
- 89090-766
- 89090-814
- 89090-842
- 89090-908
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Especificaciones
Product Family Options
Información del producto
- CapacityST Stopper No.ToleranceAvailabilityPrice
- Supplier Number: 5642-5Especificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.29619-198Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 5642-10Especificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89091-098Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 100 ml13±0.08 mlPaquete de 6$7,230.00Case de 2 (6UN)$4,682.51Supplier Number: 5642C-100Especificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89090-602Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 200 ml13±0.10 mlPaquete de 6$8,440.00Case de 2 (6UN)$16,880.00Supplier Number: 5642-200Especificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89090-676Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 1 L22±0.30 mlDiscontinued-Supplier Number: 5642-1LEspecificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89090-710Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 500 ml19±0.20 mlPaquete de 6$11,510.00Case de 2 (6UN)$14,759.53Supplier Number: 5642-500Especificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89090-748Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 250 ml16±0.12 mlDiscontinued-Supplier Number: 5642-250Especificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89090-766Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 25 ml9±0.03 mlPaquete de 6$6,950.00Case de 2 (6UN)$9,305.52Supplier Number: 5642-25Especificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89090-814Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 2 L27±0.50 mlPaquete de 1$2,960.00Case de 4 (1UN)$4,088.87Supplier Number: 5642-2LEspecificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89090-842Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 50 ml9±0.05 mlPaquete de 6$6,970.00Case de 2 (6UN)$5,255.12Supplier Number: 5642C-50Especificaciones:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89090-908Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey