PYREX® Volumetric Flasks, Class A, Low Actinic, Glass Stopper, Corning
Supplier: Corning
Certificates
About this item
PYREX®, borosilicate glass, amber, low actinic, with tapered ground stopper.
- Individually serialized
- Markings are sharp and permanent
- Low actinic glass provide protection for material sensitive to light
Each flask is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards. The protective color is an integral part of the flask, which retains its mechanical strength, chemical stability and thermal resistance.
Certifications: Calibrated to meet ASTM specifications E-694, E-542, and E-288
Specifications
- Catalog No:
- 89090-086
- 89090-088
- 89090-084
- 89090-102
- 89090-100
- 89090-094
- 89090-092
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey