Orders LinkContactUsLinkComponent
PYREX® Volumetric Flasks with [ST] Stopper, Corning
PYREX® Volumetric Flasks with [ST] Stopper, Corning
   Sustainable Options Available
Supplier:   Corning
PYREX® Volumetric Flasks with [ST] Stopper, Corning
Supplier:  Corning
   Sustainable Options Available
Certificates
PYREX® Volumetric Flasks with [ST] Stopper, Corning
PYREX® Volumetric Flasks with [ST] Stopper, Corning
3 Options Available Below
Product Details & Documents
Made of PYREX® borosilicate glass.
  • Graduation line is sharp and permanent
  • Large white block letters make inscriptions easy to read
  • Supplied with interchangeable ground glass stoppers
Flasks are designed for those that do not require the precision accuracy of our Class A capacity flasks. Capacity tolerances are twice those specified for class A volumetric ware.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 89091-108
  • 89090-822
  • 89090-916
  • 89090-608
  • 89090-754
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
  • Capacity
    ST Stopper No.
    Tolerance
    Availability
    Price
  • 89091-108
    Pyrex® Volumetric Flasks with Glass Stopper, 10 ml
    Pyrex® Volumetric Flasks with Glass Stopper, 10 ml
    10 ml
    9
    ±0.04 ml
    Log in for availability
    Log in for availability
    Case (12UN)
    Case (12UN) - $12,420.00
    $12,420.00
    Case (12UN)
       $12,420.00
    Supplier Number: 5641-10
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-108
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 89090-822
    Pyrex® Volumetric Flasks with Glass Stopper, 25 ml
    Pyrex® Volumetric Flasks with Glass Stopper, 25 ml
    25 ml
    9
    ±0.06 ml
    Discontinued
    Supplier Number: -
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-822
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 89090-916
    Pyrex® Volumetric Flasks with Glass Stopper, 50 ml
    Pyrex® Volumetric Flasks with Glass Stopper, 50 ml
    50 ml
    9
    ±0.10 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 6
    Paquete de 6 - $7,320.00
    $7,320.00
    Log in for availability
    Case de 2 (6UN)
    Case de 2 (6UN) - $8,120.32
    $8,120.32
    Paquete de 6
       $7,320.00
    Case de 2 (6UN)
       $8,120.32
    Supplier Number: 5641-50
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-916
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 89090-608
    Pyrex® Volumetric Flasks with Glass Stopper, 100 ml
    Pyrex® Volumetric Flasks with Glass Stopper, 100 ml
    100 ml
    13
    ±0.16 ml
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    89090-608
    Environmentally Preferable
    Environmental certification
  • 89090-754
    Pyrex® Volumetric Flasks with Glass Stopper, 500 ml
    Pyrex® Volumetric Flasks with Glass Stopper, 500 ml
    500 ml
    19
    ±0.40 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 6
    Paquete de 6 - $206.68
    $206.68
    Log in for availability
    Case de 2 (6UN)
    Case de 2 (6UN) - $413.36
    $413.36
    Paquete de 6
       $206.68
    Case de 2 (6UN)
       $413.36
    Supplier Number: 5641-500
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-754
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact