PYREX® Volumetric Flasks, Lifetime Red™, [ST] Stopper, Class A, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, clear, with glass stoppers.
These flasks have the capacity mark etched as a fine line through a narrow red band into clear glass.
Calibrated 'to contain' at 20°C.
Certifications: ASTM E-694, E-542, E-288
Specifications
- Catalog No:
- 89090-686
- 89090-704
- 89090-742
- 89090-810
- 89090-596
- 89090-902
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey