Orders LinkContactUsLinkComponent
PYREXPLUS® Plastic-Coated Volumetric Flasks, [ST] Joint, Class A, Corning
 :   Corning
PYREXPLUS® Plastic-Coated Volumetric Flasks, [ST] Joint, Class A, Corning
 :  Corning
PYREXPLUS® Plastic-Coated Volumetric Flasks, [ST] Joint, Class A, Corning
PYREXPLUS® Plastic-Coated Volumetric Flasks, [ST] Joint, Class A, Corning
PYREX® borosilicate glass, plastic coated, clear, with glass stoppers.
  • Resistant to thermal shock
  • Strength increased through machine-blown bodies
  • Sharp graduation line and permanent and large white block letters make the inscription easy to read
These flasks features a protective PVC coating for safety and long life, and it prevents glass from shattering and reduces spills.
Can be written on with any standard lab marker, permanent or water-soluble. Supplied with [ST] PYREX® stopper. It allows for a better hand grip and clamp hold.
Coating is autoclavable to 121°C (250°F). Do not heat above 121°C (250°F) moist heat or 110°C (230°F) dry heat.
 : ASTM E-694, E-542, E-288
 : Caution: Do not place plastic-coated ware over direct heat or open flame.
  • Catalog No:
  • 29618-751
  • 89091-674
  • 89091-692
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Product Family Options
  • Capacity
    ST Stopper No.
    Tolerance
  • 100 ml
    13
    ±0.08 ml
    Case (12UN)
    Case (12UN) - $30,570.00
    $30,570.00
    Case (12UN)
       $30,570.00
     : 65640-100
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    29618-751
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 L
    22
    ±0.30 ml
    Case (4UN)
    Case (4UN) - $12,813.06
    $12,813.06
    Case (4UN)
       $12,813.06
     : 65640-1L
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89091-674
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 500 ml
    19
    ±0.20 ml
    Case (12UN)
    Case (12UN) - $47,310.00
    $47,310.00
    Case (12UN)
       $47,310.00
     : 65640-500
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89091-692
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact