Metric/English Vernier Caliper, Mitutoyo
Catalog # 19155-057
Supplier: MITUTOYO AMERICA CORPORATION
Specifications
- Resolución:0.02 mm (0.001")
- Referencia:19155-057
- Gama de medición:0–150 mm (0–6")
- Nº de proveedor:530-312
Specifications
About this item
Made of high-caliber stainless steel caliper with etched graduations
Supplied with locking screw and soft plastic case.