Orders LinkContactUsLinkComponent
VWR® Digital Calipers
VWR® Digital Calipers
Supplier:  VWR International
Certificates
VWR® Digital Calipers
VWR® Digital Calipers
4 Options Available Below
Product Details & Documents
Calipers measure outside diameter, inside diameter, and depth.
  • Ergonomic Design
  • Traceable®
Ideal for measuring outside diameter of stoppers, inside diameter of tubing, and depth of vials. Zero button instantly sets closed jaws or any reference point to zero. Floating Zero™ may also be set in mid-measurement to read “difference results” without calculations. Calipers feature ergonomic design and durable hardened stainless steel or composite carbon fiber construction. Easy-to-read, 0.6 cm (1/4") high, digital readout switches from millimeters to inches at the touch of a button. Friction thumb roller allows for fast readings. Depth bar extends from end of instrument. No lubrication is ever required on the slide bar.
 : A serial-numbered certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. Calibrated in an ISO 17025 calibration laboratory accredited by A2LA.
 : Supplied with battery (runs for 10,000 hours).
Product Family Options
Product Information
  • Material
    Resolution
    L×W
    Accuracy
    Measurement Range
    Availability
    Price
  • Carbon fiber
    0.1 mm (0.01")
    17.8 x 6.3 cm (7 x 21/2")
    ±0.2 mm (±0.01")
    0–10.2 cm (0–4")
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,366.19
    $1,366.19
    Each (1UN)
       $1,366.19
  • Carbon fiber
    0.1 mm (0.01")
    24.1 x 7.6 cm (91/2 x 3")
    ±0.2 mm (±0.01")
    0–15.2 cm (0–6")
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,515.16
    $1,515.16
    Each (1UN)
       $1,515.16
  • Stainless steel
    0.01 mm (0.0005")
    22.9 x 7.6 cm (9 x 3")
    ±0.03 mm (±0.001")
    0–15.2 cm (0–6")
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $4,263.47
    $4,263.47
    Each (1UN)
       $4,263.47
  • Stainless steel
    0.01 mm (0.0005")
    27.9 x 7.6 cm (11 x 3")
    ±0.03 mm (±0.001")
    0–20.3 cm (0–8")
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $5,845.24
    $5,845.24
    Each (1UN)
       $5,845.24