Round-Bottom Flasks with Threaded Side Port, Ace Glass Incorporated
Catalog # 71001-548
Supplier: ACE GLASS, INC.
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacidad:250 ml
- Descripción:Heavy Wall
- Referencia:71001-548
- Center Neck [SJ] Joint:24/40
- Nº de proveedor:6936-56
- Side Port:#11 Ace-Thred
Specifications
About this item
Round-bottom flasks with [ST] outer joint center neck and Ace-Thred side port
The outer joint is tooled with reinforcement rings (full-sized joints only) for added strength and stability. The internally threaded side port can accommodate thermometers or bleed tubes. Available in heavy or standard wall. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks. Ace-Thred side port size (below) also refers to the outside diameter of tubing or thermometer that will fit this joint. Fabricated from borosilicate glass.
Ordering information: Supplied with nylon bushing and FETFE O-ring.