Orders LinkContactUsLinkComponent
Synthware Flat-Bottom Flasks, Kemtech America
Synthware Flat-Bottom Flasks, Kemtech America
500 ml, 24/40
Catalog # 60002-242
Supplier:   KEMTECH AMERICA INC
Supplier #:  F124500
Synthware Flat-Bottom Flasks, Kemtech America
Catalog # 60002-242
Synthware Flat-Bottom Flasks, Kemtech America
Synthware Flat-Bottom Flasks, Kemtech America
500 ml, 24/40
Price
$746.52
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Single-neck, flat-bottom flasks with an outer joint.
  • Single neck
  • Ground glass joint
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
  • Flat bottom
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Especificaciones
  • ST Joint No.:
    24/40
  • Capacity:
    500 ml