TRIlites® 994 Tri-Polymer Gloves, AdvanTech, Mapa Professionnel
Unique tri-polymer blend of neoprene, nitrile, and natural rubber combines comfort with added strength, tear resistance, and chemical splash protection
- Powder-free for no cross-contamination
- Rolled cuff
- Unlined
- Ambidextrous
- Color: Purple
Textured fingertips have a lightly halogenated finish for a high level of tactile sensitivity and grip in wet or dry conditions. Exclusive finish allows easy donning and glove layering, and helps keep tape and labels from sticking to gloves.
: Exceed ASTM D3578 AQL. In compliance with 21 CFR 170-199. CE marked.
: Gloves are flat-packed 100 per sealed, printed bag or dispenser box.
: This product contains natural rubber latex proteins, which may cause allergic reactions.
- Catalog No:
- 46600-162
- 46600-160
- 32916-140
- 46600-164
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
Frequently Bought Together