VWR® Disposable Gloves, PE
Use these gloves for excellent hand and product protection in virtually any type of laboratory, food handling, industrial safety, veterinary, electronics, painting, printing, or photography environment.
- Ultra soft and pliable
- Tough, waterproof construction
- Resistant to most chemicals and detergents
- Extra-long cuffs provide added protection
- Convenient pull-out box dispenser
- Ambidextrous, embossed finish
Soft, virgin polyethylene gloves are so pliable you can handle even small laboratory apparatus with ease. Resistant to most chemicals or detergents. Ambidextrous. Embossed finish.
: Gloves are made using only listed materials and levels outlined in CFR 21 170-199.
Use these gloves for excellent hand and product protection in virtually any type of laboratory, food handling, industrial safety, veterinary, electronics, painting, printing, or photography environment.
- Ultra soft and pliable
- Tough, waterproof construction
- Resistant to most chemicals and detergents
- Extra-long cuffs provide added protection
- Convenient pull-out box dispenser
- Ambidextrous, embossed finish
Soft, virgin polyethylene gloves are so pliable you can handle even small laboratory apparatus with ease. Resistant to most chemicals or detergents. Ambidextrous. Embossed finish.
: Gloves are made using only listed materials and levels outlined in CFR 21 170-199.
- Catalog No:
- 32915-224
- 32915-246
- 32915-268
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
Product Family Options
- Glove sizeGlove ColorLengthThickness
- STranslucent29.2 cm (11½")1.2 mil.-Product available on GSA Advantage®
Specifications
Cat. No.32915-224Ambidextrous / hand-specificAmbidextrous - MTranslucent29.8 cm (11¹¹⁄₁₆")1.2 mil.Paquete de 100$475.41Case de 10 (100UN)$4,761.00Product available on GSA Advantage®
Specifications
Cat. No.32915-246Ambidextrous / hand-specificAmbidextrous - LTranslucent30.5 cm (12")1.2 mil.-
Specifications
Cat. No.32915-268Ambidextrous / hand-specificAmbidextrous