Orders LinkContactUsLinkComponent
STANSOLV® Flock-Lined Nitrile Gloves, Mapa Professionnel
STANSOLV® Flock-Lined Nitrile Gloves, Mapa Professionnel
Supplier:   NEWELL BRANDS INC
STANSOLV® Flock-Lined Nitrile Gloves, Mapa Professionnel
Supplier:  NEWELL BRANDS INC
Certificates
STANSOLV® Flock-Lined Nitrile Gloves, Mapa Professionnel
STANSOLV® Flock-Lined Nitrile Gloves, Mapa Professionnel
1 Options Available Below
Product Details & Documents
Offer hand protection for industrial applications such as plating, metal finishing, chemical processing, abrasive material handling, maintenance, and acid etching.
  • Curved fingers and tapered wrist for natural, comfortable wear
  • Easy donning and doffing
  • Flock lining absorbs perspiration for added comfort
  • Flat cuffs
  • Hand-specific
Double-dipped gloves are highly resistant to aromatic petroleum and chlorinated solvents, acids, alkalies, grease, oils, and Freon®. Nitrile formulation is free of inert fillers and silicone for superior chemical resistance. Gloves contain no natural rubber proteins and will not cause allergic reactions. Also resist punctures, snags, cuts, and abrasions.
Color: Green
Certifications: In compliance with 21 CFR 170-199. CE marked.
Caution: Note: Not recommended for use with ketones, strong oxidizing acids, and organic chemicals containing nitrogen.
Specifications for Product Family
Specifications
  • Catalog No:
  • 32917-718
  • 32930-112
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
Specifications
Product Family Options
Product Information
  • Model
    Glove size
    Glove Color
    Length
    Thickness
    Availability
    Price
  • 32917-718
    Z-Pattern Grip Texture
    Z-Pattern Grip Texture
    AF-18
    8
    Green
    33 cm (13")
    18 mil.
    Discontinued
    Supplier Number: 483428ZQK
    Specifications:
    More
    Specifications
    Cat. No.
    32917-718
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Disposable
  • 32930-112
    Diamond Grip Texture
    Diamond Grip Texture
    AF-493
    10
    Green
    39.4 cm (15½")
    22 mil.
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 12
    Paquete de 12 - $3,285.15
    $3,285.15
    Log in for availability
    Case de 4 (12UN)
    Case de 4 (12UN) - $3,580.81
    $3,580.81
    Paquete de 12
       $3,285.15
    Case de 4 (12UN)
       $3,580.81
    Supplier Number: 493420
    Specifications:
    More
    Specifications
    Cat. No.
    32930-112
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Disposable