Round-Bottom Boiling Flasks with Two Necks, Heavy Wall, Chemglass
Round bottom, heavy wall, two-neck flask with [ST] outer joints on both the center neck and 20°-angled side neck
Flasks with 10/30 side neck are for use with non mercury joint thermometers and to aid in clamping of the flask, the 29/26 center neck flasks have a groove tooled just below the grind.
Both side neck and center neck have the same joint size.
: Flasks with joint sizes other than those listed are available.
Frequently Bought Together