Orders LinkContactUsLinkComponent
Evaporating Flasks, Heavy Wall, Chemglass
Supplier:   CHEMGLASS INC.
Evaporating Flasks, Heavy Wall, Chemglass
Supplier:  CHEMGLASS INC.
Certificates
Evaporating Flasks, Heavy Wall, Chemglass
Evaporating Flasks, Heavy Wall, Chemglass
2 Options Available Below
Product Details & Documents
Evaporating flask for use with rotary evaporators
Flasks have a heavy wall for vacuum applications and are balanced to prevent run-out during rotation.
Product Family Options
Uncoated Flasks
Información del producto
  • Capacity
    ST Joint
    Availability
    Price
  • 80068-964
    Single Neck Evaporating Flask
    Single Neck Evaporating Flask
    50 ml
    24/40
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,338.86
    $1,338.86
    Each (1UN)
       $1,338.86
    Supplier Number: CG-1512-01
  • 80068-980
    Single Neck Evaporating Flask
    Single Neck Evaporating Flask
    1 L
    24/40
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,558.60
    $3,558.60
    Each (1UN)
       $3,558.60
    Supplier Number: CG-1512-09