Designed for micro-electronic work where electro-static discharge (ESD) must be avoided, these comfortable industrial gloves with nitrile foam ensure that surfaces stay clean and uncontaminated. X-Static® yarn and static-reducing coating help prevent damage to sensitive electronic components and injury.
- Ensure uncontaminated surfaces
- Static-reducing coating
- Oil is channeled away for a sure grip
- Tolerance: ±0.5"
A thin design improves the handling of ever smaller components, while the ergonomic liner allows for precision in assembly applications.
Use in the aerospace, appliance manufacturing, electronics manufacturing, medical laboratory and research laboratory industries. Uses include assembling; inspecting and packing small or delicate parts; electrical component assembly and installation.
The foam nitrile-coated palm channels oil away from the glove surface for a sure grip, enabling detailed handling of small parts.
- Catalog No:
- 47727-530
- 47727-524
- Ambidiestro/Específico para cada mano:
- Específico para cada mano
- Específico para cada mano
- Desechable / Reutilizable:
- Reutilizable
- Reutilizable
- Material del revestimiento:
- Fibra conductora/nailon
- Fibra conductora/nailon
- Acabado:
- Palm Coated
- Palm Coated
- Tipo de puño:
- Puño elástico
- Puño elástico
- Normativa EN:
- 3131
- 3131