HyFlex® Foam Gloves, Ansell
8, Gray, 22.9 cm (9")
Catalog # 33890-046
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Supplier #: 103332
Jump to:
Product Details & Documents
These gloves provide comfort and dexterity when working with small, oily parts.
- Hand-specific
- Excellent abrasion resistance
- Elastic cuffs
The nitrile coating helps reduce hand fatigue and channels oil away from the glove surface, providing a surer grip. Fine-gauge, ultra-lightweight knit liner offers excellent resistance to nicks and abrasions.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Glove Color:Gray
- Glove size:8
- Length:22.9 cm (9")
- Cat. No.:33890-046
- Ambidextrous / hand-specific:Hand-specific