KONTES® Round-Bottom Boiling Flasks, Vertical Type, Necks, Heavy Wall, Kimble Chase
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificados
Acerca de este artículo
The tops of all three necks are approximately the same height, with Standard Taper outer joints.
- Side necks are vertical
Designed from ASTM Specification E1378, Type IV requirements. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Certificaciones: Designed from ASTM Specification E1378, Type IV requirements.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Información de pedido: For additional volumes and [ST] joints, contact your Avantor sales representative.
Frequently Bought Together