KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
Jump to:
Acerca de este artículo
All flasks have a short neck and a full length [ST] 24/40 interchangeable ground glass joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
3 Options Available Below
Product Details & Documents
All flasks have a short neck and a full length [ST] 24/40 interchangeable ground glass joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Documents
Certificates
Buscar por número de loteKimble Petrochemical Glassware Brochure
Documentación del producto
Product Family Options
Información del producto
- CapacityST Joint No.O.D.×HAvailabilityPrice
- 125 ml24/4065 x 115 mmDiscontinued-Supplier Number: 25055-125
- 250 ml24/4083 x 133 mmPaquete de 1$881.07Case de 12 (1UN)$10,572.84Supplier Number: 25055-250
- 500 ml24/40102 x 155 mmPaquete de 1$842.75Case de 12 (1UN)$10,113.02Supplier Number: 25055-500
- 1 L24/40130 x 182 mmPaquete de 1$1,003.65Case de 12 (1UN)$12,043.84Supplier Number: 25055-1000