Orders LinkContactUsLinkComponent
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
Certificates
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
1 Options Available Below
Product Details & Documents
Short neck flasks are designed to ASTM Specification E1403, Class 3 requirements
They feature a full-length, outer, [ST] 24/40 joint.
Documents
Product Family Options
Product Information
  • Capacity
    ST Joint No.
    O.D.×H
    Availability
    Price
  • 100 ml
    24/40
    60 x 115 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 2
    Paquete de 2 - $1,417.18
    $1,417.18
    Log in for availability
    Case de 6 (2UN)
    Case de 6 (2UN) - $8,503.05
    $8,503.05
    Paquete de 2
       $1,417.18
    Case de 6 (2UN)
       $8,503.05
    Supplier Number: 25285-100
    Specifications:
    More
    Specifications
    Cat. No.
    89001-282
  • 3 L
    24/40
    185 x 255 mm
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    89001-294