UltraSense™ EC Powder-Free Nitrile Gloves, Microflex®, Ansell
Proveedor: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificados
Acerca de este artículo
Ultra-soft, powder-free gloves are made from a nitrile formulation that has the fit and feel of latex and provides reliable protection against many hazardous and infectious substances.
- Ambidextrous
- Beaded cuffs
- Textured fingers
Non-stick surface resists tapes and adhesives.
Precaución: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Especificaciones
- Catalog No:
- 89173-616
- 89065-284
- 89065-280
- 89065-282
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Desechable
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Espesor del puño:
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- Espesor del dedo:
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- Espesor de la palma:
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil