MidKnight™ Nitrile Examination Gloves, Microflex®, Ansell
Supplier: MICROFLEX CORPORATION MX
Certificates
About this item
Powder-free MidKnight™ Nitrile Examination gloves provide exceptional tactile sensitivity, comfort, and durability.
- Powder-free, fully textured for firm grip
- Exceptional tactile sensitivity, comfort, and durability
- Distinct color masks stains
- Thickness: 5.5 mil (finger)
Distinct color masks stains. Fully textured for a firm grip.
Caution: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Specifications
- Catalog No:
- 94001-370
- 94001-366
- 94001-368
- 94001-364
- 94001-372
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Espesor del puño:
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- Espesor del dedo:
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- Espesor de la palma:
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
Specifications
Related Information
Frequently Bought Together