PYREX® Weighing Bottles, Low Form, Cap Stopper, Corning
Catálogo # 77776-964
Proveedor: Corning
Product available on GSA Advantage®
Especificaciones
- [ST] unión No.:55/12
- Capacidad:35 ml (1.2 oz.)
- Dimensiones:50 O.D. x 30 H, mm (115/16 x 13/16")
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
- Referencia:77776-964
- Peso:68 g (2.4 oz.)
- Nº de proveedor:1688-5030
Especificaciones
Acerca de este artículo
PYREX® borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
These low form weighing bottles have an external standard taper joint style opening. The closure style cap fits over the externally ground body.