PYREX® Round Bottom Kjeldahl Flask with Long Neck, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, long neck.
- Have tooled finished necks for extra strength and uniform stopper fit
- Optimum balance between thermal shock and chemical resistance
- Can be used in ASTM E-258 procedure
Suitable for the determination of nitrogen. Care must be taken to ensure the bottoms of the flasks do not come in contact with the heating elements. Kjeldahl flask have ring supports for this purpose.
Specifications
- Catalog No:
- 89090-476
- 89091-336
- 89091-316
- 89091-426
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey