Orders LinkContactUsLinkComponent
Nalgene® Filtering Flasks, Polypropylene, Thermo Scientific
Nalgene® Filtering Flasks, Polypropylene, Thermo Scientific
Nalgene® Filtering Flasks, Polypropylene, Thermo Scientific
Certificates
Nalgene® Filtering Flasks, Polypropylene, Thermo Scientific
Nalgene® Filtering Flasks, Polypropylene, Thermo Scientific
1 Options Available Below
Product Details & Documents
One-piece molded for strength and stability
Withstand full vacuum to 762mm Hg (30in. Hg). Tubulation at 45° angle prevents tipping. Oval marking area. Barbed adapter accepts 6.4mm (1/4") to 9.5mm (3/8") I.D. tubing. Autoclavable.
Product Family Options
Product Information
  • Capacity
    Rubber Stopper Size
    Availability
    Price
  • 2 L
    10
    Log in for availability
    Log in for availability
    Case (1UN)
    Case (1UN) - $1,650.00
    $1,650.00
    Case (1UN)
       $1,650.00
    Supplier Number: DS4101-2000
  • 500 ml
    7
    Discontinued
    Supplier Number: -