Orders LinkContactUsLinkComponent
PYREX® Weighing Bottles, Glass, Corning
PYREX® Weighing Bottles, Glass, Corning
   Opciones sostenibles disponibles
Supplier:   Corning
PYREX® Weighing Bottles, Glass, Corning
Supplier:  Corning
   Opciones sostenibles disponibles
Certificates
PYREX® Weighing Bottles, Glass, Corning
2 Options Available Below
Product Details & Documents
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
These tall or low form bottles have a standard taper joint style opening that accepts a closed bottom, hollow pennyhead style stopper.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 77776-934
  • 77776-946
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Especificaciones
Product Family Options
Tall Form
Información del producto
  • Capacity
    Dimensions
    ST Joint No.
    Weight
    Availability
    Price
  • 16 ml (0.54 oz.)
    25 O.D. x 50 H, mm (1 x 2")
    24/12
    21 g (0.7 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (24UN)
    Case (24UN) - $19,890.00
    $19,890.00
    Case (24UN)
       $19,890.00
    Supplier Number: 1680-2550
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    77776-934
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
Low Form
Información del producto
  • Capacity
    Dimensions
    ST Joint No.
    Weight
    Availability
    Price
  • 50 ml (1.7 oz.)
    60 O.D. x 30 H, mm (23/8 x 13/16")
    60/12
    71 g (2.5 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (6UN)
    Case (6UN) - $11,760.00
    $11,760.00
    Case (6UN)
       $11,760.00
    Supplier Number: 1682-6030
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    77776-946
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact