PYREX® Erlenmeyer Flasks, Wide Mouth, Corning
Supplier: Corning
Certificates
About this item
Flasks made of PYREX® borosilicate glass, graduated.
- Used as a vented culture flask when optional membrane cap is used
- Convenient single size plug-seal cap
- One-piece autoclavable orange polypropylene plug seal caps and drip-free pouring rings
For preparing and storing nutrient media and to use for larger volume serological pipets and ideal for use as a shaker flask.
Specifications
- Catalog No:
- 32645-038
- 32645-044
- 32645-046
- 32645-048
- 32645-052
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Specifications
Related Information
Frequently Bought Together