PYREX® Erlenmeyer Flasks, Graduated, Narrow Mouth, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Heavy duty rim flasks have approximate graduations in durable white enamel. Extra large marking space. Heavy, durable top tooling.
Specifications
- Catalog No:
- 29136-106
- 29136-048
- 29136-060
- 29136-026
- 29136-037
- 89090-858
- 89090-738
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Specifications
Related Information
Frequently Bought Together