PYREX® Erlenmeyer Flasks, Graduated, [ST] Stopper, Corning
Proveedor: Corning
Certificados
Acerca de este artículo
PYREX® borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These narrow mouth have standard taper stoppers for secure sealing. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance. For convenience, these flasks are graduated to show approximate capacity. The 125 ml flasks are satisfactory for use as weighing bottles.
Especificaciones
- Catalog No:
- 89090-650
- 89090-788
- 89091-454
- 89090-876
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey