PYREX® Griffin Beakers, Graduated
89090-524
: Corning
: 1000-4L
Product available on GSA Advantage®
- Catalog No:
- 89090-524
- 89090-522
- 13912-069
- 13912-284
- 13912-127
- 13912-091
- 13912-320
- 470211-310
- 470211-332
- 13912-149
- 470211-314
- 470211-326
- 13912-182
- 470211-328
- 13912-160
- 470211-330
- 13912-229
- 13912-207
- 470211-320
- 13912-240
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
PYREX® Griffin Beakers are made of PYREX® borosilicate glass.
- Very good chemical resistance
- Thermal shock resistance
PYREX® Griffin Beakers are manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength.
Beakers come in sizes 250 ml and larger have a double graduated metric scale to indicate approximate content. The 10 ml size is not graduated. All sizes have an extra large marking spot.
: Meet ASTM E960 specifications.
: The variety pack includes one of each of the following beakers: 50, 100, 250, 600, and 1000 ml.
Frequently Bought Together