DURAN® Storage/Media Bottles, Glass, with Caps, SI Analytics
Catálogo # 29110-902
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Tamaño de la tapa:45 mm
- Capacidad:1 L (33.8 oz.)
- Dimensiones:23.0 x 10.1 cm (9 x 4")
- Referencia:29110-902
- Nº de proveedor:218015455
Specifications
About this item
Ideal for storage, mixing, and sampling.
- Resistant to chemical attack and thermal shock
- All components are autoclavable
These borosilicate glass bottles feature white graduation marks in milliliter increments and a marking spot.
Delivery: Supplied with blue polypropylene screw caps and pouring rings.