ContactUsLinkComponent
Formic Acid/Acetonitrile Blend 0,1% (v/v) LabReady™ LC-MS para LC-MS
Flammable
Irritant
Formic Acid/Acetonitrile Blend 0,1% (v/v) LabReady™ LC-MS para LC-MS
Catalog # BJLC441-1
Product available on GSA Advantage®
CAS Number:  
Flammable
Irritant
Formic Acid/Acetonitrile Blend 0,1% (v/v) LabReady™ LC-MS para LC-MS
Catalog # BJLC441-1
Supplier Number:  LC441-1
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Tipo de env.:
    Glass bottle, GL45
  • Tamaño:
    1 L
  • Referencia:
    BJLC441-1
  • Nº de proveedor:
    LC441-1
Specifications
About this item
 :75-05-8
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:–46 °C
Density:0.7767 g/cm³ (25 °C)
Flash Pt:8 °C
Storage Temperature:Ambiente
EINECS:200-579-1
UN:1648
ADR:3,II
 :GHS02
 :GHS07