Protector® Nitrile/Natural Rubber Gloves, MAPA Professional®
Hand protection for a wide variety of applications including chemical processing, acid cleaning, construction, food processing, metal machining, and agricultural chemicals.
- Curved fingers and hand-specific design for natural fit
- Embossed safety grip
Soft cotton flock-lined natural rubber gloves with flat cuff. Resistant to acids, alkalies, cutting oils, alcohols, and grease. Nitrile blend over-dip in work area provides superior chemical, puncture, and wear resistance, and immediate visible indication of wear. Color: green with blue over-dip.
Note: Not recommended for use with ketones or strong oxidizing acids and organic chemicals containing nitrogen.
Certifications: Gloves are made using only listed materials and levels outlined in 21CFR, Section 177.2600.
- Glove Color:Blue
- Glove size:M
- Length:33 cm (13")
- Thickness:28 mil.
- Cat. No.:32930-056
- Disposable / reusable:Disposable
- Ambidextrous / hand-specific:Ambidextrous