Nalgene® Erlenmeyer Flasks with Polypropylene Screw Cap, Polycarbonate, Thermo Scientific
Proveedor: Thermo Fisher Scientific
Certificados
Acerca de este artículo
Wide-mouth flasks feature excellent clarity and mechanical strength, making them ideal for culture work.
Polypropylene closure provides dependable seal for use with liquid culture media. Both flasks and caps are autoclavable. Before autoclaving, set cap on top of flask without engaging threads.
Frequently Bought Together