Orders LinkContactUsLinkComponent
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific
Wide-mouth flasks are built with heavy walls for hard use and a wide base for stability.
  • Liquid level easily visible
  • Autoclavable
The 2L flask has molded-in graduations.
Product Family Options
  • Capacity
    Stopper No.
  • 125 ml
    3
    Paquete de 12
    Paquete de 12 - $4,280.00
    $4,280.00
    Case de 2 (12UN)
    Case de 2 (12UN) - $5,096.64
    $5,096.64
    Paquete de 12
       $4,280.00
    Case de 2 (12UN)
       $5,096.64
     : 4102-0125
  • 250 ml
    6
    Paquete de 6
    Paquete de 6 - $2,710.00
    $2,710.00
    Case de 2 (6UN)
    Case de 2 (6UN) - $3,222.01
    $3,222.01
    Paquete de 6
       $2,710.00
    Case de 2 (6UN)
       $3,222.01
     : 4102-0250
  • 1000 ml
    8
    Paquete de 2
    Paquete de 2 - $2,020.00
    $2,020.00
    Case de 3 (2UN)
    Case de 3 (2UN) - $2,401.86
    $2,401.86
    Paquete de 2
       $2,020.00
    Case de 3 (2UN)
       $2,401.86
     : 4102-1000
  • 2000 ml
    10
    Paquete de 2
    Paquete de 2 - $2,510.00
    $2,510.00
    Case de 3 (2UN)
    Case de 3 (2UN) - $2,864.22
    $2,864.22
    Paquete de 2
       $2,510.00
    Case de 3 (2UN)
       $2,864.22
     : 4102-2000