- Color del guante:Green
- Tamaño del guante:9
- Ambidiestro/Específico para cada mano:Específico para cada mano
- Desechable / Reutilizable:Desechable
- Con polvo/sin polvo:Sin polvo
- Longitud:33 cm (13")
- Grosor:11 mil.
- Referencia:32917-858
- Nº de proveedor:519319
STANSOLV® A-10CR gloves provide hand protection against solvents and other chemicals that attack natural rubber. Ideal for solvent-based photo-resists, caustic developer solutions, and organic-based strippers. Used for semiconductor wet processes using solvent-based strippers, etchants, and cleaners. Also used for handling cleanroom and clean process acids, caustics, and solvents.
- Case-hardened finish for added chemical resistance
- Easy on/off
Gloves resist organic solvents such as xylene, perchloroethylene, freon, ethylene glycol ether, and Cellosolve®
Hand-specific gloves are powder-free with flat cuffs.
Cut-, puncture-, and abrasion-resistant. Unique embossed Z-pattern grip improves wet gripping. Finished, tested, and packaged in Class 100 cleanroom and 100% inspected for leaks. Gloves contain no natural rubber proteins and will not cause allergic reactions.