Orders LinkContactUsLinkComponent
PYREX® Erlenmeyer Flasks, Graduated, Screw Cap, Corning
 :   Corning
PYREX® Erlenmeyer Flasks, Graduated, Screw Cap, Corning
 :  Corning
PYREX® Erlenmeyer Flasks, Graduated, Screw Cap, Corning
PYREX® Erlenmeyer Flasks, Graduated, Screw Cap, Corning
Made of PYREX® borosilicate glass, with phenolic screw caps.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Graduated to show approximate capacity
  • With marking spot
Erlenmeyer flasks with phenolic screw caps are ideal for use as a shaker flask or for mixing, media prep and storage needs. Impact resistant caps are equipped with cemented-in, inert white rubber liners which are resistant to steam sterilization. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance.
  • Catalog No:
  • 89090-648
  • 89090-786
  • 89090-870
  • 89090-874
  • 89090-922
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Product Family Options
  • Capacity
    Graduation Interval
    Graduation Range
    Thread
  • 125 ml
    25 ml
    50 - 125 ml
    24 - 410
    Paquete de 12
    Paquete de 12 - $10,150.00
    $10,150.00
    Case de 2 (12UN)
    Case de 2 (12UN) - $13,068.21
    $13,068.21
    Paquete de 12
       $10,150.00
    Case de 2 (12UN)
       $13,068.21
     : 4985-125
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89090-648
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 250 ml
    25 ml
    50 - 200 ml
    38 - 430M
    Paquete de 6
    Paquete de 6 - $5,430.00
    $5,430.00
    Case de 4 (6UN)
    Case de 4 (6UN) - $8,072.17
    $8,072.17
    Paquete de 6
       $5,430.00
    Case de 4 (6UN)
       $8,072.17
     : 4985-250
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89090-786
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1000 ml
    50 ml
    250 - 1000 ml
    38 - 430M
     : 4985-1L
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89090-870
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 500 ml
    50 ml
    100 - 500 ml
    38 - 430M
    Paquete de 6
    Paquete de 6 - $5,500.00
    $5,500.00
    Case de 4 (6UN)
    Case de 4 (6UN) - $10,844.93
    $10,844.93
    Paquete de 6
       $5,500.00
    Case de 4 (6UN)
       $10,844.93
     : 4985-500
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89090-874
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 50 ml
    10 ml
    20 - 50 ml
    24 - 410
    Paquete de 12
    Paquete de 12 - $10,270.00
    $10,270.00
    Case de 2 (12UN)
    Case de 2 (12UN) - $11,159.65
    $11,159.65
    Paquete de 12
       $10,270.00
    Case de 2 (12UN)
       $11,159.65
     : 4985-50
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89090-922
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact