PYREXPLUS® Media Bottles, Graduated, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, with screw cap.
- Resistant to thermal shock
- Teal enameled graduations and marking spot
- Glass bead indicates full capacity line
These heavy duty bottles are ideal for storage, mixing and sampling. A protective PVC coating prevents glass from shattering and reduces spills. It includes green colored PP plug seal cap with drip free pouring rings.
Autoclavable to 121 °C (249 °F) and have thread size: GL45
Caution: Do not tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. Do not place bottles over direct heat or flame. Do not heat above 121 °C (249 °F) moist heat or 110 °C (230 °F) dry heat.
Specifications
- Catalog No:
- 16157-057
- Environmentally Preferable:
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
Specifications
Related Information
Frequently Bought Together