KIMTECH PURE® G3 White Nitrile Gloves, Kimberly-Clark
Supplier: Ansell Healthcare
Certificates
About this item
Powder-free nitrile gloves have textured fingertips to facilitate control and grip.
- Static dissipative in use
- Ambidextrous
- Beaded cuff
Certifications: Recommended for ISO Class 4 (FED-STD-209E Class 1/M1.5) or higher cleanroom environments. AQL 1.5.
Ordering information: Packaged in a Class 100 cleanroom. Packaged in cleanroom-compatible polyethylene, 100 gloves per double poly bag.
Specifications
- Catalog No:
- 14215-764
- 14215-768
- 14215-762
- 14215-760
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Espesor del dedo:
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- Espesor de la palma:
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil