PURPLE NITRILE™ and PURPLE NITRILE-XTRA™ Examination Gloves, Kimberly-Clark
Nitrile gloves offer excellent tactile sensitivity and dexterity as well as superior fit and comfort.
- Offer excellent tactile sensitivity and dexterity
- Textured fingertips and beaded cuff
- Ambidextrous and powder-free
- Length: 24.1 cm (9.5") or 30.5 cm (12")
- Thickness: 5.9 mil (finger), 4.7 mil (palm), 3.5 mil (cuff)
Beaded cuff for added strength and easier donning. Glove fingers are thicker for greater barrier protection. Textured fingertips enhance wet and dry grip. Ambidextrous and powder-free. Color: purple.
: Exceed ASTM standard for critical defects. Meets Chemotherapy Permeation Testing per ASTM D6978-05.
: Packaged in dispenser boxes.
- Catalog No:
- 32934-076
- 32934-080
- 32933-968
- 32934-078
- 32934-084
- 32933-972
- 32934-082
- 32933-970
- 32933-974
- 32933-976
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Espesor del dedo:
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- Espesor de la palma:
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
Frequently Bought Together