TouchNTuff® 92-675 Nitrile Gloves, Disposable, Ansell
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificates
About this item
Ideal for manufacturing, light-duty maintenance and cleanup, intricate parts handling, lab analysis/technical work, and automotive applications.
- Rolled cuffs
- Ambidextrous
- Durable, yet soft for improved comfort
- Powder-free
- Static-dissipative
Textured fingers for exceptional grip.
Certifications: Component materials comply with FDA regulations for food contact.
Specifications
- Catalog No:
- 32889-884
- 32889-888
- 32889-890
- 32889-886
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Desechable
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Acabado:
- Textured
- Textured
- Textured
- Textured
- Tipo de puño:
- Rolled, beaded
- Rolled, beaded
- Rolled, beaded
- Rolled, beaded
Specifications
Related Information
Certificates
Search by Lot NumberFocus Safety Volume 1 Issue 1 Safety Essentials Volume 2
Product Literature
Frequently Bought Together