Acerca de este artículo
Ambidextrous, powder-free gloves with beaded cuffs for strength and reduction of cuff roll down.
- Superior strength and puncture resistance
- Certificates of analysis available
- Quality data available
Nitrile is superior in strength and puncture resistance compared to other glove compounds. Film thickness designed for comfort and tactile sensitivity. Gloves are 6.3 mil thick at fingers and 5.1 mil thick at palms. Textured fingertips for a reliable grip. Uncompromising manufacturing processes yield consistently low particle counts. Proprietary nitrile, silicone-free formulation, and specialized manufacturing processes result in consistently low levels of ionic extractables and nonvolatile residue (NVR) transfer. In-use static dissipative properties superior to natural rubber latex.
The 30.5 cm (12") length and tapered cuff provide more coverage over the garment sleeve.
Especificaciones
- Catalog No:
- 40101-358
- 40101-356
- 40101-353
- 40101-352
- 40101-357
- 40101-354
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Espesor del dedo:
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- Espesor de la palma:
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil