Microflex® FreeForm® EC FFE-775 Examination Gloves, Ansell Healthcare, Ansell
Proveedor: MICROFLEX CORPORATION MX
Certificados
Acerca de este artículo
Extended protection and grip, approved for use by first responders.
- Non-stick properties resists tape and adhesives
When performing mild-duty work, MICROFLEX® FreeForm® EC is a long cuff nitrile glove that offers reliable protection and a secure grip. Its extended cuff provides expanded protection over the wrist and forearm.
Certificaciones: ASTM D6319, EN 420:2003 + A1:2009, FDA21 CFR 177-2600, NFPA 2012, US QSR/FDA 510(k) Medical Examination Grade
Embalaje: Gloves are packaged 50 per dispenser, 10 dispensers per case (500 gloves per case).
Precaución: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Especificaciones
- Catalog No:
- 32916-780
- 32916-778
- 32916-782
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Latex/No contiene Latex:
- No contiene Latex
- No contiene Latex
- No contiene Latex
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Acabado:
- Textured fingers
- Textured fingers
- Textured fingers
- Tipo de puño:
- Extended
- Extended
- Extended
- Espesor del puño:
- 3.5 mil
- 3.5 mil
- 3.5 mil
- Espesor del dedo:
- 0.16 mm (6.3 mil)
- 0.16 mm (6.3 mil)
- 0.16 mm (6.3 mil)
- Espesor de la palma:
- 0.12 mm (4.7 mil)
- 0.12 mm (4.7 mil)
- 0.12 mm (4.7 mil)