Nitrile Gloves, Kimtech™ G3 NXT™, Kimberly-Clark
These powder-free nitrile gloves feature textured fingertips to facilitate control and grip.
- Powder free
- Textured fingertips for control and grip
- Ambidextrous
- Beaded cuff
AQL 1.5.
: Recommended for ISO Class 4 (FED-STD-209E Class 1/M1.5) or higher cleanroom environments.
: Packaged in a Class 100 cleanroom. Packed 100 gloves per double poly bag with case liner.
- Catalog No:
- 14214-540
- 14214-542
- 14214-538
- 14214-536
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Espesor del dedo:
- 6.2 mil
- 6.2 mil
- 6.2 mil
- 6.2 mil
- Espesor de la palma:
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
Frequently Bought Together