Orders LinkContactUsLinkComponent
CE5 System Nitrile Gloves, Microflex®, Ansell
CE5 System Nitrile Gloves, Microflex®, Ansell
CE5 System Nitrile Gloves, Microflex®, Ansell
CE5 System Nitrile Gloves, Microflex®, Ansell
These ambidextrous gloves are for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) controlled environments.
  • Silicone-free
  • Powder-free
  • Superb tactile sense and comfort
  • Low surface particulates
These gloves produce only very low levels of surface particulates, ionic extractables, and ono-volatile residues.
 : Lot-specific Certificate of Analysis available upon request.
 : Packed 100 gloves per vacuum-sealed double poly bag, 10 poly bags per case.
 : Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
  • Catalog No:
  • 32916-762
  • 32916-764
  • 32916-766
  • 32916-768
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finger thickness:
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • Palm thickness:
  • 3.9 mil
  • 3.9 mil
  • 3.9 mil
  • 3.9 mil
Product Family Options
  • Glove size
    Glove Color
    Length
  • S
    White
    30.5 cm (12")
    Case (1,000UN)
    Case (1,000UN) - $9,510.00
    $9,510.00
    Case (1,000UN)
       $9,510.00
     : CE5-755-S
    Specifications
    Cat. No.
    32916-762
    Ambidextrous / hand-specific
    Ambidextrous
    Powdered / powder-free
    Powder-free
    Finger thickness
    4.7 mil
    Palm thickness
    3.9 mil
  • M
    White
    30.5 cm (12")
    Case (1,000UN)
    Case (1,000UN) - $9,510.00
    $9,510.00
    Case (1,000UN)
       $9,510.00
     : CE5-755-M
    Specifications
    Cat. No.
    32916-764
    Ambidextrous / hand-specific
    Ambidextrous
    Powdered / powder-free
    Powder-free
    Finger thickness
    4.7 mil
    Palm thickness
    3.9 mil
  • L
    White
    30.5 cm (12")
    Case (1,000UN)
    Case (1,000UN) - $9,510.00
    $9,510.00
    Case (1,000UN)
       $9,510.00
     : CE5-755-L
    Specifications
    Cat. No.
    32916-766
    Ambidextrous / hand-specific
    Ambidextrous
    Powdered / powder-free
    Powder-free
    Finger thickness
    4.7 mil
    Palm thickness
    3.9 mil
  • XL
    White
    30.5 cm (12")
    Case (1,000UN)
    Case (1,000UN) - $9,510.00
    $9,510.00
    Case (1,000UN)
       $9,510.00
     : CE5-755-XL
    Specifications
    Cat. No.
    32916-768
    Ambidextrous / hand-specific
    Ambidextrous
    Powdered / powder-free
    Powder-free
    Finger thickness
    4.7 mil
    Palm thickness
    3.9 mil