Nitrilite® 93-311, Nitrile Gloves, Ansell
Specifically designed for strength and comfort in critical environments, Nitrilite 93-311 significantly reduces particulate and extractable levels.
- Strength and comfort in critical environments
- Significantly reduces particulate and extractable levels
It is 100% powder-free with no plasicizers, silicone, or pigments.
Ideal for electronics manufacturing, medical laboratories, pharmaceutical, and research laboratories.
Its nitrile material is thinner than comparable vinyl or latex gloves for a lighter, more comfortable glove with improved dexterity and tactility. Ideal applications include electronics assembly, handling sensitive parts and materials, pharmaceutical manufacturing, product protection in handling: cosmetics packaging , optical goods , microelectronic and micro-mechanic components, and protection from blood and other bodily fluids.
: Manufactured in ISO 9002 registered facilities. Each lot is tested, certified, and documented. AQL for pinholes: 4.0.
: Gloves feature cleanroom-compatible packaging containing 50 gloves per inner poly bag and 2 inner bags per outer poly bag. Lot number is printed on outer poly bag in IPA-resistant ink.
- Catalog No:
- 47748-210
- 47748-216
- 47748-214
- 47748-212
- 47748-218
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Sin polvo
- Espesor del dedo:
- 5 mil
- 5 mil
- 5 mil
- 5 mil
- 5 mil
- Espesor de la palma:
- 4 mil
- 4 mil
- 4 mil
- 4 mil
- 4 mil
Frequently Bought Together