Synetron® Polymer-Coated Latex Examination Gloves, Microflex®, Ansell
Gloves are made from soft natural rubber latex for improved elasticity and greater comfort.
- Interior polymer coating for superior antisticking properties in all conditions
- Fully textured, beaded cuff
- Powder-free
Ambidextrous gloves are fully textured to provide a superior grip in wet and dry conditions.
: Packaged 50 gloves per dispenser box.
: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 32916-542
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Latex/No contiene Latex:
- Latex
- Con polvo/sin polvo:
- Sin polvo
- Espesor del puño:
- 6.7 mil
- Espesor del dedo:
- 11.8 mil
- Espesor de la palma:
- 9.1 mil
Frequently Bought Together